missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Fine Pore Cassettes

For very small biopsy tissue samples, cassettes have a front-edge lid hinge with attached lids that are available in 12 different color varieties

349.00 €

Specifications

Closure Included Yes
Packaging Type Loose Packed
Quantity 1000/Cs.
Pore Style Fine Pore
PrintMate Compatible No
View More Specs

Products 12
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
16364086
View Documents
Epredia™
B851735BE
Beige
349.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16374086
View Documents
Epredia™
B851735BL
Blue
349.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16384086
View Documents
Epredia™
B851735GN
Green
349.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16394086
View Documents
Epredia™
B851735GY
Gray
349.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16304096
View Documents
Epredia™
B851735LA
Lavender
349.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16314096
View Documents
Epredia™
B851735LG
Light Green
349.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16324096
View Documents
Epredia™
B851735LI
Lilac
349.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16334096
View Documents
Epredia™
B851735OR
Orange
349.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16344096
View Documents
Epredia™
B851735PK
Pink
349.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16354096
View Documents
Epredia™
B851735TN
Tan
349.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16364096
View Documents
Epredia™
B851735WH
White
349.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
16374096
View Documents
Epredia™
B851735YW
Yellow
349.00 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing, Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving
Specifications

Specifications

Yes
1000/Cs.
No
Loose Packed
Fine Pore
Cassette
Videos
SDS
Documents

Documents

Product Certifications