missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Fine Pore Cassettes in Tube Packs
For use with very small biopsy tissue samples. These cassettes are compatible with all Epredia PrintMate™ Cassette Printers, have a front-edge lid hinge, and attached lids that are available in 12 color varieties.
287.00 €
Specifications
Closure Included | Yes |
---|---|
Quantity | 750/Cs. |
Pore Style | Fine Pore |
Closure Type | Attached Lid, Front Hinge |
PrintMate Compatible | Yes |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
16344076
|
Epredia™
B851734BE |
Beige |
287.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16354076
|
Epredia™
B851734BL |
Blue |
287.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16364076
|
Epredia™
B851734GN |
Green |
287.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16374076
|
Epredia™
B851734GY |
Gray |
287.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16384076
|
Epredia™
B851734LA |
Lavender |
287.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16394076
|
Epredia™
B851734LG |
Light Green |
287.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16304086
|
Epredia™
B851734LI |
Lilac |
287.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16314086
|
Epredia™
B851734OR |
Orange |
287.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16324086
|
Epredia™
B851734PK |
Pink |
287.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16334086
|
Epredia™
B851734TN |
Tan |
287.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16344086
|
Epredia™
B851734WH |
White |
287.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16354086
|
Epredia™
B851734YW |
Yellow |
287.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
Yes | |
Fine Pore | |
Yes | |
Cassette |
750/Cs. | |
Attached Lid, Front Hinge | |
For Use With PrintMate™ Printers |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ Fine Pore Cassettes in Tube Packs