missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ Fine Tip LDPE Pasteur Pipette

Pasteur pipette for simple and safe liquid measurement and transfer.

Brand:  Wheaton™ 711117F10S

 View more versions of this product

Product Code. 16424999

  • 250.00 € / Pack of 1500
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For liquid transfer
  • Designed to ensure safe sample transfer
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Not Autoclavable
No
LDPE
20 μL
Bulb
102 mm
1 mL
CE Marked
Sterile
No
10/Pk
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.