missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Blue PVC glove.
- Cotton flocked interior.
- Diamond-patterned finish for grasping dry and damp objects.
- Outstanding flexibility and dexterity.
- Tailored for multi-purpose and repetitive handling in an oily atmosphere.
- Low resistance to ketones, aromatic and halogen hydrocarbons.
- Long cuff to protect the forearm.

Specifications
Specifications
Certifications/Compliance | CE CATEGORY II Intermediary risks, EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards, EN 374-2003 Chemical risks and against micro-organisms, EC Category PPE 3, EC Certificate Number 0075/797/162/01/09/0051, EC Attestation Number 0075/797/162/01/09/0051 |
Classification | Dexterity: 5 |
Color | Blue |
Laboratory/Cleanroom Use | Yes |
Material | PVC, Cotton |
Product Type | Gloves |
Size | 7 |
Thickness (Metric) | 0.5 to 1 mm |
For Use With (Application) | Building, Construction, Catering, Chemical, Fishing, Food, Industrial Cleaning, Laboratory, Medical, Pharmaceutical, Printing, Ship Building and Water Treatment industries. |
Liner Material | Cotton flock |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Honeywell Safety Products™ Finedex™ PVC Soft Gloves >
Spot an opportunity for improvement?Share a Content Correction