Learn More
Thermo Scientific™ Finger Tite Connectors
Ensure reliable, leak-free connections for optimal GC performance using these Finger Tite Connectors.
Brand:  Thermo Scientific™ 290ST132
 View more versions of this product
Product Code. 10321485
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Ensure reliable, leak-free connections for optimal GC performance using Thermo Scientific™ Finger Tite Connectors. The perfect metal-to-metal seal eliminates contamination and produces lower air/water background noise compared to conventional graphite/polymer ferrules. They are easy to handle even with a hot injector/detector and do not require the use of tools. Unlike other ferrules, they do not require retightening after installation.
Recommended Use
  • Thermo Scientific™ and Agilent™ GC instruments
  • Connectors for SSL and MS systems
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Split/Splitless and FID Starter Kit
Injector/Detector Starter Kit
TRACE GC Ultra and FOCUS GC Instruments
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.