Learn More
Thermo Scientific™ Finnpipette™ F1 GLP Kits
Access everything needed for pipetting in these all-inclusive kits featuring state-of-the-art pipetters that maximize reliability and comfort.
Brand: Thermo Scientific™ 4700860
Description
Access everything needed for pipetting in these all-inclusive kits. Thermo Scientific™ Finnpipette™ F1 GLP (Good Laboratory Pipetting) Kit includes state-of-the-art Finnpipette F1 pipetters which offer superior performance and unique features such as adjustable finger rest and antimicrobial surface for user comfort.
Features:
- Adjustable finger rest for increased ergonomicss and comfortability
- Set and forget pipetting button for easy and secured volume adjustment
- Antimicrobial surface for better protection
- Large easy to read display for better vision ergonomics
- Soft-touch tip ejection for light end effortless tip ejection
Kit 1 includes:
- Three Thermo Scientific Finnpipette Pipetters from 1 to 1000μL:
- Finnpipette F1, 1-10μL
- Finnpipette F1, 10-100μL
- Finnpipette F1, 100-1000μL
- Finntip Flex tips: 10, 1x96; 200, 1x96; 1000, 1x96
- F-stand; F1 pen; F1 brochure
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Kit 2 includes:
- Four Thermo Scientific Finnpipette Pipetters from 0.2 to 1000μL:
- Finnpipette F1, 0.2-2μL
- Finnpipette F1, 2-20μL univ
- Finnpipette F1, 20-200μL
- Finnpipette F1, 100-1000μL
- Finntip Flex tips: 10, 1x96; 200, 2x96; 1000, 1x96
- F-stand; F1 pen; F1 brochure
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Kit 3 includes:
- Three Thermo Scientific Finnpipette Pipetters from 10 to 10000μL:
- Finnpipette F1, 10-100μL
- Finnpipette F1, 100-1000μL
- Finnpipette F1, 1000-10000μL
- Finntip Flex tips: 200, 1x96; 1000, 1x96; FT 10mL, 1x24 F-stand; F1 pen; F1 brochure
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Kit 4 includes:
- Three Thermo Scientific Finnpipette Pipetters from 10 to 1000μL:
- Finnpipette F1, 2-20μL
- Finnpipette F1, 20-200μL
- Finnpipette F1, 100-1000μL
- Flex tips: (2) 200μL, 10x96; 100-1000μL, 10x96
- F-stand; F1 pen; F1 brochure
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Ordering Information:
Good Laboratory Pipetting kit also includes samples of Finntip Flex for maximum performance and best ergonomics. The kit contains a Good Laboratory Pipetting guide offering a extensive information of pipetting and factors influencing to the pipetting results.
Compatible with:
Finntip, ART tip, HLT tip
Specifications
Finntip Flex tips: 10; 200; 1000 | |
Assorted | |
Finnpipette™ F1 | |
Finnpipette™ F1 | |
0.2 to 1000 μL |
Kit 2: Finnpipettes F1 0.2-2μL micro, 2-20μL, 20-200μL and 100-1000μL | |
Finntip: Flex Tips: 10, 200, 1000 | |
1 | |
GLP Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.