missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Finnpipette™ F1 Good Laboratory Pipetting (GLP) Kits
GREENER_CHOICE

Enjoy the convenience of having a variety of Finnpipette™ F1 single channels packaged together, in a range of volumes, with compatible Finntip pipette tips and a pipette stand to support good laboratory pipetting in your lab

970.00 € - 1180.00 €

Specifications

Number of Channels 1
Type GLP Kit
View More Specs

Products 4
Product Code Brand Description Volume (Metric) Compatible Tips Price Quantity & Availability  
Product Code Brand Description Volume (Metric) Compatible Tips Price Quantity & Availability  
15541945
GREENER_CHOICE
View Documents
Thermo Scientific™
4700850N
Finnpipette F1 GLP Kit 1: 1-10μL micro, 10-100μL and 100-1000μL 1 to 1000 μL Finntip Flex tips: 10; 200; 1000
970.00 €
Each
In Stock
to see stock.
Add to basket
 
15551945
GREENER_CHOICE
View Documents
Thermo Scientific™
4700860N
Finnpipette F1 GLP Kit 2: 0.2-2μL 2-20μL, 20-200μL and 100-1000μL 0.2 to 1000 μL Finntip Flex tips: 10; 200; 1000
1180.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
15561945
GREENER_CHOICE
View Documents
Thermo Scientific™
4700865N
Finnpipette F1GLP Kit 3: 10-100μL, 100-1000μL and 1-10mL 10 to 10,000 μL Finntip Flex tips: 200; 1000; FinnTip 10mL
985.00 €
Each
In Stock
to see stock.
Add to basket
 
15571945
GREENER_CHOICE
View Documents
Thermo Scientific™
4701060N
Finnpipette F1 GLP Kit 4: 2-20μL, 20-200μL and 100-1000μL 2 to 1000 μL Finntip Flex tips: 200; 1000μL
970.00 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

Support good laboratory pipetting in your lab with the Thermo Scientific™ Finnpipette™ F1 GLP Kits, which offer a variety of single channels in a range of volumes that are packaged with compatible Finntips, and a convenient pipette stand. Select the kit that is right for you, and take advantage of the Finnpipette F1 performance and lightweight design to maximize your comfort.

For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.

Finnpipette F1: Everything you want in your everyday, go-to pipette

Features:

  • Lightweight handle design
  • Low plunger and tip attachment & ejection forces
  • Secure volume lock to prevent undesirable volume drift during operation
  • Silent, effortless volume adjustment
  • Adjustable finger rest - comfort for both right- and left-handed users
  • Color coded together with Finntips for easier recognition of compatible tips

Kit 1 includes:

  • Three Thermo Scientific Finnpipette F1 Pipettes from 1 to 1000 μL:
    • 1-10 μL, micro
    • 10-100 μL
    • 100-1000 μL
  • Grip Strip for each handle to improve grip
  • Flex tips: 10 micro: 1 x 96; 200: 1 x 96; 1000: 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 2 includes:

  • Four Thermo Scientific Finnpipette F1 Pipettes from 0.2 to 1000 μL:
    • 0.2-2 μL
    • 2-20 μL univ
    • 20-200 μL
    • 100-1000 μL
  • Grip Strip for each handle to improve grip
  • Flex tips: 10 micro: 1 x 96; 200: 2 x 96; 1000: 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 3 includes:

  • Three Thermo Scientific Finnpipette F1 Pipettes from 10 to 10,000 μL:
    • 10-100 μL
    • 100-1000 μL
    • 1000-10,000 μL
  • Grip Strip for each handle to improve grip
  • Flex tips: 200: 1 x 96; 1000: 1 x 96; FT 10 mL: 1 x 24
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 4 includes:

  • Three Thermo Scientific Finnpipette F1 Pipettes from 20 to 1000 μL:
    • 2-20 μL
    • 20-200 μL
    • 100-1000 μL 
  • Grip Strip for each handle to improve grip
  • Flex tips: 200μL: 2 x 96; 1000 μL: 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Ordering Information:

The kit contains samples racks of Finntip Flex, the most advanced traditional tip offering very low attachment and ejection forces. The Good Laboratory Pipetting Guide contains helpful tips for pipetting including pipetting techniques, ergonomic considerations and how to select tips for best performance.

Warranty: 5 years with web registration.

"

Specifications

Specifications

1
GLP Kit
Videos
SDS
Documents

Documents

Product Certifications