Learn More
Thermo Scientific™ Finnpipette™ F1 Multichannel Pipettes
GREENER_CHOICE
Get optimal performance and ergonomic comfort from these F1 Multichannel Pipettes. Available in 8-, 12- or 16-channel models; choose from a vast selection of Finntip pipette tips for optimal results.
Brand:  Thermo Scientific™ 4661080N
977.80 EUR valid until 2025-12-31
Use promo code "24315" to get your promotional price.
 View more versions of this product
Product Code. 15541935
  • 1235.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Show More
Specifications
Specifications
Finntip 20 Micro, Filter 20 Micro
Purple
1 to 10 μL
Purple
Finnpipette F1
16
0.02μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Finnpipette™ F1 Multichannel Pipettes > Finnpipette™ F1 16-Channel Pipette, Volume range: 1 to 10μL; Color Code: Purple; Compatible tips: Finntip 20; Unit Size: 1 piece