Learn More
Thermo Scientific™ Finnpipette™ F1 Variable Volume Pipettes
GREENER_CHOICE
Maximize user comfort and reduce strain with the F1 system, which delivers performance with precision, all in a lightweight design. It's the ideal system for applications using expensive reagents and valuable samples.
Brand:  Thermo Scientific™ 4641030N
245.44 EUR valid until 2025-12-31
Use promo code "24314" to get your promotional price.
 View more versions of this product
Product Code. 15551885
  • 386.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Show More
Specifications
Specifications
Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
Pink
Variable Volume Pipette
0.02μL
Finnpipette F1
1
1 to 10 μL
Pink
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Finnpipette™ F1 Variable Volume Pipettes > Finnpipette™ F1 Pipette, Volume range: 0.2 to 2μL; Color Code: Pink; Compatible tips: Finntip Flex 10 and Finntip 10; Unit Size: 1 piece