missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Finnpipette™ F2 Good Laboratory Pipetting (GLP) Kits
GREENER_CHOICE

Enjoy the convenience of having a variety of Finnpipette™ F2 single channels packaged together, in a range of volumes, with compatible Finntip pipette tips and a pipette stand to support good laboratory pipetting in your lab

930.00 € - 1350.00 €

Specifications

Autoclavable Autoclavable
Color Assorted
Model F2
Product Line Finnpipette F2
Type GLP Kit
View More Specs

Products 4
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11835850
GREENER_CHOICE
View Documents
Thermo Scientific™
4700870
Finnpipette F2 GLP Kit 1: 1-10μL micro, 10-100μL and 100-1000μL
1080.00 €
1 set
In Stock
to see stock.
Edge
Add to basket
 
11845850
GREENER_CHOICE
View Documents
Thermo Scientific™
4700880
Finnpipette F2 GLP Kit 2: 0.2-2μL micro, 2-20μL, 20-200μL and 100-1000μL
1350.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
11855850
GREENER_CHOICE
View Documents
Thermo Scientific™
4700885
Finnpipette F2 GLP Kit 3: 10-100μL, 100-1000μL and 1-10mL
1005.00 €
1 set
In Stock
to see stock.
Add to basket
 
11773027
GREENER_CHOICE
View Documents
Thermo Scientific™
4701070
Finnpipette F2 GLP Kit 4: 2-20μL, 20-200μL and 100-1000μL
930.00 €
1 set
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Access everything needed for pipetting in these all-inclusive kits. Thermo Scientific™ Finnpipette™ F2 GLP (Good Laboratory Pipetting) Kits provide durable, high-performance Finnpipette F2 pipettes which offer maximum reliability and comfort. Built with tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light, these variable volume pipettes are fully autoclavable without disassembling.

For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.

Finnpipette F2: Proven reliable for long-term intensive use

Features:

  • Fully autoclavable for protection
  • Tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light
  • Easy maintenance
  • Color coded together with Finntips for easier recognition of compatible tips

Kit 1 includes:

  • Three Thermo Scientific Finnpipette F2 Pipettes from 1 to 1000 μL:
    • 1-10 μL micro
    • 10-100 μL
    • 100-1000 μL
  • Flex tips: 10 micro: 1 x 96; 200: 1 x 96; 1000: 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 2 includes:

  • Four Thermo Scientific Finnpipette F2 Pipettes from 0.2 to 1000 μL:
    • 0.2-2 μL
    • 2-20 μL univ
    • 20-200 μL
    • 100-1000 μL
  • Flex tips: 10 micro: 1 x 96; 200: 2 x 96; 1000: 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 3 includes:

  • Three Thermo Scientific Finnpipette F2 Pipettes from 10 to 10,000 μL:
    • 10-100 μL
    • 100-1000 μL
    • 1000-10,000 μL
  • Flex tips: 200: 1 x 96; 1000: 1 x 96; FT 10 mL: 1 x 24
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 4 includes:

  • Three Thermo Scientific Finnpipette F2 Pipettes from 20 to 1000 μL:
    • 2-20 μL
    • 20-200 μL
    • 100-1000 μL 
  • Flex tips: 200: 2 x 96; 1000: 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Ordering Information:

The kit contains samples of Finntip Flex, the most advanced traditional tip offering very low attachment and ejection forces. The Good Laboratory Pipetting Guide contains helpful tips for pipetting including pipetting techniques, ergonomic considerations and how to select tips for best performance.

Warranty: 5 years with web registration.

Specifications

Specifications

Autoclavable
F2
GLP Kit
Assorted
Finnpipette F2
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.