Learn More
Thermo Scientific™ Finnpipette™ Novus GLP Kits
Access everything needed for pipetting in these all-inclusive GLP kits featuring ergonomic and easy-to-use pipetters.
1410.00 € - 1760.00 €
Product Code | Brand | Description | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Includes | Price | Quantity & Availability | ||||
11859133
|
Thermo Scientific™
4700470 |
Finnpipette Novus GLP Kit 1 | Novus 100 and 1000µL single channel pipettes, two Novus stands, two sample Finntip racks |
1410.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11869133
|
Thermo Scientific™
4700490 |
Finnpipette Novus GLP Kit 2 | Novus 1000 and 5000µL single channel pipettes, two Novus stands, two sample Finntip racks |
1410.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11485668
|
Thermo Scientific™
4701120 |
Finnpipette Novus GLP Kit 3 | Novus 10micro, 100 and 1000µL single channel pipettes, three Novus stands, three sample Finntip racks |
1760.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11495668
|
Thermo Scientific™
4701130 |
Finnpipette Novus GLP Kit 4 | Novus 100, 1000 and 10000µL single channel pipettes, three Novus stands, three sample Finntip racks |
1760.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Access everything needed for pipetting in these all-inclusive Thermo Scientific™ Finnpipette™ Novus GLP kits. The kits cover the volume range you need with four different options to choose from, and include Finnpipette™ Novus™ pipetters, appropriate stands and sample tips.
Novus GLP Kit 1 includes:
- Finnpipette Novus 10-100μL
- Finnpipette Novus 100-1000μL
- Two Finnpipette Novus stands
- One rack of Finntip Flex 200 and Flex 1000
- Reagent Reservoirs demo pack
- Finntip Sample Pack
- Pipetting Guide
- Finnpipette Novus brochure
Novus GLP Kit 2 includes:
- Finnpipette Novus 100-1000μL
- Finnpipette Novus 0.5-5mL
- Two Finnpipette Novus stands
- One rack of Finntip 5ml and Flex 1000
- Reagent Reservoirs demo pack
- Finntip Sample Pack
- Finnpipette Novus brochure
- Pipetting Guide
Novus GLP Kit 3 includes:
- Novus pipette, 1-10μL, micro
- Novus pipette, 10-100μL
- Novus pipette, 100-1000μL
- Three Finnpipette Novus pipette stands
- One rack of Flex 10, Flex 200 and Flex 1000
- Reagent Reservoirs demo pack
- Finntip Sample Pack
- Pipetting Guide
- Novus brochure
Novus GLP Kit 4 includes:
- Novus pipette, 10-100μL
- Novus pipette, 100-1000μL
- Novus pipette, 1-10mL
- Three Finnpipette Novus pipette stands
- One rack of Flex 200, Flex 1000 and Finntip 10ml
- Reagent Reservoirs demo pack
- Finntip Sample Pack
- Pipetting Guide
- Novus brochure
Specifications
1 | |
GLP Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.