Learn More
Thermo Scientific™ Finnpipette™ Novus i Multichannel Electronic Pipette
Product Code. 15102589
Click to view available options
Includes:
Pipette Stand Only
Pipetter, Pipette Stand
Unit Size:
Each
Product Code. 15102589
Brand: Thermo Scientific™ 991S
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Combine these pipettes with our adjustable pipette stand to assist in lower throughput MSIA sample enrichment.
Combine Thermo Scientific™ Finnpipette™ Novus i Multichannel Electronic pipettes with our adjustable pipette stand for an economical semi-automated system for rapid development and application of Mass Spectrometric Immunoassay (MSIA) methods. Each Novus i comes with a custom MSIA menu for further simplification of pipette programming that is complementary to MSIA applications.
Features:
  • Simplify MSIA methods using Finnpipette Novus i pipette on the pipette stand
  • New software that allows 1-999 aspirate and dispense cycles with one press on the pipette trigger
  • Continuous battery charging of pipette on the adjustable stand during sample processing
  • Include forward, reverse, diluting, stepper pipetting, mixing, sequential stepping and more; Nine aspirate/dispense speeds
Compatible with:
Thermo Scientific MSIA D.A.R.T.'S™ (ordered separately)
TRUSTED_SUSTAINABILITY
Specifications
Description Finnpipette Novus i adjustable pipetter stand
Product Type Pipette Stand
Includes Pipette Stand Only
Product Line Finnpipette Novus i