Learn More
Thermo Scientific™ Finnpipette™ Stands
GREENER_CHOICE
Product Code. p-4520840
Click to view available options
Product Type:
Carousel Stand Adapter
Carousel Stand with Cover
Pipette Stand
Shelf Hanger
Holds:
1
3
Electronic Pipette
Manual Pipette
Manual Pipettes (3)
Manual Pipettes (6)
Manual Pipettes (6) or Electronic Pipettes (3)
Multi-channel Manual Pipette
Unit Size:
1 set
Each
Product Code. 11455748
Brand: Thermo Scientific™ 9420400
Only null left
Add to basket
Add to basket
Provide safe and convenient storage for manual and electronic pipettes using these stands, with options for single stand, F-stand, multi-stand, multichannel and carousel.
Provide safe and convenient storage for manual and electronic pipettes using Thermo Scientific™ Finnpipette™ Stands. Available options include options for single stand, F-stand, multi-stand, multichannel and carousel.
Finnpipette F-Stand
  • Accommodates six pipettes
  • Sturdy crossbar structure with slot for each pipette for secure storage
  • Suitable for both single channel and multichannel models
Finnpipette Multichannel Stand
  • Multi-position stand
  • Can hold any multichannel Finnpipette
  • Pipette can be stored in upright or horizontal position
  • Small footprint
Finnpipette Carousel Stand
  • Rotating, portable
  • Transparent lid protects pipettes from contamination
  • Shipped with three adapters for electronic Finnpipette Novus pipettes and five adapters for manual Finnpipette pipettes
Finnpipette Novus Stand
  • For convenient storage of electronic Finnpipette Novus pipette
  • Accommodates both single and multichannel Finnpipette Novus models with ease
TRUSTED_SUSTAINABILITY
Specifications
Product Type Pipette Stand
Description F-Stand, Linear Stand, white
Color White
Holds Manual Pipettes (6)
Positions 6