missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Finntip™ Stepper Pipette Tips
GREENER_CHOICE

Product Code. 10401891
Click to view available options
Volume (Metric):
500 μL
1250 μL
2.5 mL
5 mL
12.5 mL
25 mL
50 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 10
Pack of 100
Pack of 20
Pack of 25
Pack of 50
This item is not returnable. View return policy

Product Code. 10401891

Brand: Thermo Scientific™ 9404220

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Integrate Thermo ScientificTM FinntipTM Stepper Pipette Tips with the Thermo ScientificTM FinnpipetteTM Stepper Pipetter.

Integrate Thermo Scientific™ Finntip™ Stepper Pipette Tips with the Thermo Scientific™ Finnpipette™ Stepper Pipetter.

  • Nonsterile tips are bulk packaged; sterile tips are individually wrapped
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 25 mL
Volume (Metric) Dispensing 500, 1000, 1500, 2000, 2500 μL
Filtered Non-filtered
Sterility Non-sterile
Certifications/Compliance ISO 9001, ISO 14001
Format Bulk
For Use With (Equipment) Thermo Scientific FinnPipette Stepper
Product Line FinnTip
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.