Learn More
DWK Life Sciences Fiolax™ Borosilicate Test Tube, with beaded rim
Test tubes are universal cylindrical laboratory glassware with a round bottom that are used e.g. for chemical reactions, pretreatments or for storing small amounts of liquid.
26.00 € - 97.95 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10599841
|
DWK Life Sciences
261103603 |
80 mL |
49.55 €
Pack of 50 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
10058180
|
DWK Life Sciences
261100606 |
6 mL |
26.85 €
Pack of 100 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
10333751
|
DWK Life Sciences
261102307 |
35 mL |
51.15 €
Pack of 100 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
10682401
|
DWK Life Sciences
261102607 |
39 mL |
58.30 €
Pack of 100 |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
10091863
|
DWK Life Sciences
261100306 |
4 mL |
26.85 €
Pack of 100 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||||
10313561
|
DWK Life Sciences
261100109 |
2 mL |
26.00 €
Pack of 100 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
10282001
|
DWK Life Sciences
261102101 |
25 mL |
41.90 €
Pack of 100 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
10078190
|
DWK Life Sciences
261101302 |
16 mL |
36.90 €
Pack of 100 |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
10333801
|
DWK Life Sciences
261103303 |
60 mL |
43.75 €
Pack of 50 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
10777381
|
DWK Life Sciences
261100803 |
6.5 mL |
28.50 €
Pack of 100 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
10221961
|
DWK Life Sciences
261101602 |
20 mL |
44.84 €
Pack of 100 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
10757571
|
DWK Life Sciences
261103809 |
110 mL |
97.95 €
Pack of 50 |
In Stock
Log in to see stock. |
|||||
Description
Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
- Thin-walled and therefore resistant to rapid temperature changes or localized heating
- Easier pouring due to beaded rim
- Fired-on, highly durable white ceramic
Specifications
Fiolax | |
Autoclavable |