Learn More
3M™ Fish Protein Rapid Kit L25FSH
Intended for screening for the presence of fish proteins in clean-in-place (CIP) final rinse water, environmental swab samples, food ingredients and processed food products.
Brand: 3M™ 7100151335
This item is not returnable.
View return policy
Description
- Results visible at 11 ± 1 minute after application of the sample.
- Rapid, qualitative immunochromatographic lateral flow test cartridges designed for the detection of allergenic food proteins.
- Designed to detect both processed and unprocessed allergen proteins.
- Test can accommodate environmental swab, CIP rinse water and food samples.
Specifications
Microbiology Media | |
Allergen | |
25/Cs. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
3M™ Fish Protein Rapid Kit L25FSH
Spot an opportunity for improvement?Share a Content Correction