missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Bovine Serum Albumin (BSA) Protease-free Powder
Purified by a heat shock process
Brand: Fisher BioReagents™ BP9703-100
This item is not returnable.
View return policy
Description
CAS: 9048-46-8
Specifications
BSA Protease-free | |
Proprietary heat shock process | |
Bovine |
6.8 to 7.2 | |
100g | |
Powder |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Bovine Serum Albumin, Heat Shock, Reagent Grade Powder, pH 7.0
Signal Word
- Warning
Hazard Category
- Specific target organ toxicity Category 3
Hazard Statement
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.