missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Bovine Serum Albumin, Heat Shock Treated

Suitable for immunological studies

Brand:  Fisher BioReagents™ BP1600-1


 View more versions of this product

Product Code. 11403833

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

CAS: 9048-46-8

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Immunological Applications
7 ±0.3
Bovine
BSA, Fraction V, Heat Shock Treated
1kg
Poly Bottle
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Bovine Serum Albumin, Heat Shock, Reagent Grade Powder, pH 7.0

Signal Word
  • Warning

Hazard Category
  • Specific target organ toxicity Category 3

Hazard Statement
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers