Learn More
Fisher BioReagents™ EZ-Run™ Prestained Rec Protein Ladder, Fisher BioReagents
Comprised of 10 recombinant proteins covalently coupled to a blue chromophore plus 10 and 72kDa reference bands tagged with green and orange dyes, respectively
Brand: Fisher BioReagents™ BP3603-500
139.99 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Comprised of 10 recombinant proteins covalently coupled to a blue chromophore plus 10 and 72kDa reference bands tagged with green and orange dyes, respectively
- Ladder is ideal for monitoring protein separation during SDS-PAGE, verifying Western blot transfer efficiency and approximating the molecular weight of blotted proteins
- Supplied in gel loading buffer
The covalently coupled chromophore affects protein mobility; this prestained marker should only be used for approximating the molecular weight of separated proteins.

Specifications
Blue | |
Liquid | |
Monitoring protein separation during SDS-PAGE | |
10-170 kDa | |
100 Loadings |
Protein Molecular Weight Marker/Ladder | |
Supplied in gel-loading buffer | |
500 μL, 100 Loadings | |
−20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
This product is manufactured under the license for Strep-tag™ technology covered by U.S. Patent No. 5,506,121; U.K. Patent No. 2272698; and French Patent No. 93 13 066.