Learn More
Granulated Agar, Fisher BioReagents
Used as a solidifying agent to provide the solid medium to support the growth of microbes
Brand: Fisher BioReagents BP9744-5
This item is not returnable.
View return policy
Description
- Granulated agar media CAS:[9002-18-0]
- Store at room temperature
Specifications
Granulated Powder | |
Used as a solidifying agent to provide the solid medium to support the growth of microbes | |
5 kg |
Agar | |
Poly Pail |
Safety and Handling
Product Identifier
- Agar, Granulated