missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Microbiology Media: LB Broth, Lennox (Granulated)
Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.
Brand: Fisher BioReagents™ BP9722-500
This item is not returnable.
View return policy
Description
- Directions for Media Preparation: Add 20g to 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water) then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
- Provides a nutrient-rich environment to enable faster growth of E. coli
- Format: Granulated powder

Specifications
Granulated Powder | |
Microbiology Media | |
For maintenance and propagation of Escherichia coli in molecular biology procedures | |
500 g |
LB Broth, Lennox | |
Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 5g/L | |
Poly Bottle |
Safety and Handling
Product Identifier
- LB Broth, (Lennox L Broth)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction