missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Microbiology Media: LB Broth (Powder) - Lennox
Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.
Brand: Fisher BioReagents™ BP1427-2
This item is not returnable.
View return policy
Description
- Directions for Media Preparation: Suspend 20g in 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat to completely dissolve. Before use, sterilize by autoclaving for 15 minutes
- Provides a nutrient-rich environment to enable faster growth of E. coli
- Format: Dehydrated powder
Specifications
Bacterial Culture | |
Molecular genetics | |
Microbiology Media | |
Preparing bacterial nutrient media | |
2000 g |
Powder | |
LB Broth - Lennox | |
Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 5g/L | |
Poly Pail |
Safety and Handling
Product Identifier
- Broth Miller
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction