missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Peptone (Granulated)
Brand: Fisher BioReagents™ BP9725-500
This item is not returnable.
View return policy
Description
- Mixture of peptides and free amino acids from pancreatic hydrolysis of animal tissue
- pH (at 25°C, 2% Solution): 6.5 to 7.5
- CAS: 73049-73-7

Specifications
Granulated Powder | |
Peptone | |
Total Nitrogen <10%, Amino Nitrogen <3.5% | |
500 g |
Molecular genetics | |
Peptone | |
Poly Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction