missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Agar, Powder / Flakes, Fisher BioReagents™

Used as a solidifying agent for culture media.

93.55 €

Specifications

Grade Molecular genetics
Product Type Agar
For Use With (Application) Used for a solidifying agent for culture media
View More Specs

Products 1
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
10572775
View Documents
Fisher BioReagents™
BP1423-500
500 g Poly Bottle
93.55 €
500g
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Used as a solidifying agent for culture media

  • Used as a solidfying agent to provide the solid medium to support the growth of microbes for cell culture applications
  • Functionally tested for promotion of growth of E. coli and S. aureus
  • Typical concentration is 1.5% agar (15g/L) in water
  • CAS: 9002-18-0
  • Molecular Weight: 336.337 g/mol
  • Chemical Formula: C14H24O9
Specifications

Specifications

Molecular genetics
Agar
Used for a solidifying agent for culture media
Videos
SDS
Documents

Documents

Product Certifications