Learn More
Fisher BioReagents™ Proteinase K (Tritirachium album/Molecular Biology)
Brand: Fisher BioReagents™ BP1700-500
299.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Proteinase K (Tritirachium album/Molecular Biology)
- Not on TSCA inventory: for research and development use only; not for manufacturing or commercial purposes
Proteinase K is widely used in the isolation of DNA and RNA from cell and tissue preparations. It is a wide-spectrum protease, inactivating many enzymes, including endogenous nucleases.

Specifications
White | |
-20°C | |
500 mg | |
Solid |
Isolation of DNA and RNA from cell and tissue preparations, inactivating many Enzymes, including Endogenous Nucleases | |
Tritirachium album | |
Proteinase K |
Safety and Handling
- Proteinase K
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.