missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Tryptone (Granulated)

An enzymatic digest of casein, a milk protein

112.62 € - 344.97 €

Specifications

Form Granulated Powder
Media Type Tryptone
Product Type Microbiology Media
Formulation Total Nitrogen ≥10%, Amino Nitrogen ≥3.5%
View More Specs

Products 2
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
11385982
View Documents
Fisher BioReagents™
BP9726-2
2 kg Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11365982
View Documents
Fisher BioReagents™
BP9726-500
500 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • pH (at 25°C, 2% Solution): 6.5 to 7.3

Specifications

Specifications

Granulated Powder
Tryptone
Microbiology Media
Total Nitrogen ≥10%, Amino Nitrogen ≥3.5%
Videos
SDS
Documents

Documents

Product Certifications