missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher Chemical™ Permount™ Mounting Medium
For both mounting and long-term storage of slides
Brand: Fisher Chemical™ SP15-100
Alert:
This chemical is considered hazardous by the 2012 OSHA Hazard Communic. Standard (29 CFR 1910.1200).
Description
For both mounting and long-term storage of slides
- Low viscosity
- Gives a thinner adhesive layer for better optical quality
- A refractive index near that of fixed protein helps keep image free of optical imperfections such as diffraction halos
- Suitable for mounting cover slips to slides with thick or thin specimens
- Won't shift the colors of biological dyes
- Won't lose its cohesiveness, adhesive quality, or color with age
- Preserves most biological dyes with little or no fading when slides are stored in darkness
- Refractive index, 1.518
Specifications
Mounting and long-term storage of slides | |
Slide Mounting Media | |
100 mL |
3.38 oz. | |
Amber Glass |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Permount Mounting Media
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Toluene, Polyterpenehydrocarbon resin