missing translation for 'onlineSavingsMsg'
Learn More

Fisher Chemical™ Permount™ Mounting Medium

For both mounting and long-term storage of slides

Brand:  Fisher Chemical™ SP15-500


 View more versions of this product

Product Code. 15832544

  • 90.80 € / 500mL
Estimated Shipment: 25-06-2025
to see stock.
Explore more special offers
Add to basket

Alert:

This chemical is considered hazardous by the 2012 OSHA Hazard Communic. Standard (29 CFR 1910.1200).

This item is not returnable. View return policy

Description

Description

For both mounting and long-term storage of slides

 

  • Low viscosity
  • Gives a thinner adhesive layer for better optical quality
  • A refractive index near that of fixed protein helps keep image free of optical imperfections such as diffraction halos
  • Suitable for mounting cover slips to slides with thick or thin specimens
  • Won't shift the colors of biological dyes
  • Won't lose its cohesiveness, adhesive quality, or color with age
  • Preserves most biological dyes with little or no fading when slides are stored in darkness

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mounting and long-term storage of slides
Slide Mounting Media
500 mL
16.9 oz.
Amber Glass
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Permount Mounting Media

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : Toluene, Polyterpenehydrocarbon resin

SDS
Documents

Documents

Certificates
Special Offers

Special Offers