missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 300 Memory Water-Resistant Stopwatch

Memory storage for 300 lap times. Water-Resistant Stopwatch has an O-ring-sealed and shock-resistant ABS plastic case with nonslip rubber sides.

Brand:  Fisherbrand™ 90023-97

Product Code. 11709825


Description

Description

  • Times to 10 hours with a resolution of 0.01 of a second and accuracy of 0.001%
  • Provides stopwatch functions: single-action, time-in/-out, continuous-timing, cumulative-split, interval-split, 300 memories and repetitive programmable countdown alarm times
  • Three-line display simultaneously shows running time, cumulative-split and interval-split
  • Unit recalls 300 memories and displays sample numbers one through 300
  • Provides repeat functions of countdown alarm timing
  • Features also include functions of pacer, stroke and speed calculations
  • Displays time of day in hours, minutes and seconds as well as AM/PM, date and day of the week
  • Dimensions: 3 x 2.5 x 0.88 in. (7.6 x 6.4 x 2.2 cm)

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.

Specifications

Specifications

Water-resistant Stopwatch
0.001%
LCD
Silver Oxide
3 in.
2.12 in.
0.87 in.
2.5 oz.
2 Year Silver Oxide Always-on Battery, Lanyard, Velcro™ Fasteners for vertical mounting, and Detachable Clip
ISO 17025, A2LA, NIST
10 hr.
0.01 sec.
ABS Plastic
3 x 2.12 x 0.87 in. (1.62 x 5.38 x 2.20 cm)
1.62 cm
5.38 cm
2.20 cm
0.07 kg
Provides repeat functions of countdown alarm timing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers