Learn More
Fisherbrand™ accuflex™ Electrode Support
For accumet™ benchtop meters
Brand:  Fisherbrand™ 01X321801
Product Code. 11510234
  • 111.00 € / Each
Estimated Shipment: 17-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For accumet™ benchtop meters
  • Attaches to meter base with bracket supplied
  • Holds one micro electrode and three standard electrodes
  • Permits full up/down and left/right positioning of electrodes
  • Made of chemical-resistant plastic
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Support Arm and Bracket
pH
Electrode support arm bracket for meter
Plastic
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ accuflex™ Electrode Support > Accuflex Electrode Support Arm