Learn More
Fisherbrand™ accumet™ Automatic Temperature Compensation Probes - Mercury - Free
For accumet™ benchtop meters.
Brand:  Fisherbrand™ 01X306701
 View more versions of this product
Product Code. 11530234
  • 345.00 € / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with accumet Meters
  • Signal solution temperature to meter for automatic compensation
  • Permit temperature readout with meters having this feature
  • Probes for accumet AP Series portable meters have waterproof connector
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Accumet benchtop and portable meter
ATC
0.9 m
Stainless Steel
0.25 in. Phone