missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ Glass Body Calomel Reference Electrodes
Brand: Fisherbrand™ 93X218898
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Description
- Standard-size glass body
- Contains mercury
- -5° to +80°C
- 0 to 14pH range
Specifications
Pin | |
0 to 14 | |
Mercury | |
Hg/Hg2Cl2 (Calomel) | |
23°F to 176°F | |
5.75 in. | |
14.6 cm | |
0.37 in. | |
1.6 cm |
Glass | |
-5°C to +80°C | |
Biomedical, Tris | |
Calomel | |
80°C Max. | |
30 in. | |
76.2 cm | |
1 cm |