missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ Waterproof AP72 pH/mV/Temperature Meter
Shop All Fisherbrand Products
Click to view available options
Range (pH):
-2 to 16
Unit Size:
Each
Description
Includes
Includes includes meter, batteries and instructions.
- Ergonomic design fits comfortably in the hand
- Electrode bracket positions electrode for one-handed operation
- Advanced set-up mode lets user customize meter parameters and check electrode slope and offset to assure electrode accuracy
- Read pH, absolute mV or relative mV and temperature
- Accuracy of ±0.01pH and ±0.02mV
- Manual or automatic temperature control
- Five-point push-button pH calibration and automatic buffer recognition (pH 1.68, 4.01, 7.01, 10.01, 12.45)
- HOLD function, Ready indicator and automatic shut-off function
- Read pH, mV or relative mV and temperature in °C or °F
- Memory allows storage and recall of up to 50 readings with corresponding temperatures
Real-time clock that stamps stored data and calibration data with date and time
Six-point push-button pH calibration and automatic buffer recognition
Selectable buffer sets (NIST™, DIN and Standard US)
Certifications
ISO 9001:2000, CE
Warranty
Warranted to the end user to be free from defects in materials and workmanship under normal use for a period of three years.
Specifications
Specifications
| Includes | Meter, batteries and instructions |
| Accuracy (Temperature) | ±0.5°C |
| Accuracy (mV) | ±0.2 from 0 to ±400, ±2 beyond 200 |
| Accuracy (pH) | ±0.01 |
| For Use With (Application) | pH, mV, Temperature |
| Range (pH) | -2 to 16 |
| Resolution (pH) | 0.001 |
| Range (mV) | 0 to ±2000 |
| Resolution (mV) | 0.1 from 0 to ±400, 1 beyondmV |
| Calibration Points | 5 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur