Learn More
Fisherbrand™ Baume Hydrometers
Baume heavy scale for working with liquids heavier than water.
25.55 € - 25.60 €
Product Code | Brand | Certifications/Compliance | Increments | Length (Metric) | Baume Range | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Certifications/Compliance | Increments | Length (Metric) | Baume Range | Price | Quantity & Availability | |||
11796923
|
Fisherbrand™
11571B |
Without 3-point certification | 0.1° | 305 mm | 9 to 21° |
25.55 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||
11706933
|
Fisherbrand™
11571C |
Without 3-point certification | 0.1° | 310 mm | 19 to 31° |
25.60 €
Each |
Estimated Shipment: 05-06-2025
Log in to see stock. |
|||
Description
Baume heavy scale for working with liquids heavier than water.
- Permanently marked with individual serial numbers
Order Info
Use Fisherbrand cylinder 08-530J with 250mL of liquid for submersion, available separately
Certifications
A certificate is provided to indicate traceability to standards provided by NIST™.
Compliance
Accuracy meets or exceeds ASTM™ E 100 specifications, Calibrated according to ASTM E 126 specifications.
Specifications
Baume Hydrometer |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.