Learn More
Fisherbrand™ Dissolved Oxygen Membrane Kit
For use with Fisherbrand™ accumet™ Benchtop Dissolved Oxygen Meters
Brand:  Fisherbrand™ 01X241604
167.04 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Product Code. 11510264
  • 212.00 € / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with Fisherbrand™ accumet™ Benchtop Dissolved Oxygen Meters
  • Includes six membrane caps, polishing disk and electrolyte filling solution
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Accumet™ Benchtop Dissolved Oxygen Meter
Membrane Kit
Six membrane caps, polishing disk and electrolyte filling solution
DO