missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Frontier™ 5306 Mini Centrifuge Accessory, Adapters
For use with Frontier 5306 Mini Centrifuge
Brand: OHAUS™ 30129561
This item is not returnable.
View return policy
Description
- 0.2 mL Centrifuge Tube Adapter
- Holds up to 8 tubes
- 0.2 mL Centrifuge Tube Adapter
- Holds up to 8 tubes
Specifications
Adapter | |
Frontier 5306 mini centrifuge | |
0.2 mL | |
0/1 V |
8 x 0.2 mL | |
Adapter, 0.2ml Tube x8, FC5306 | |
8 Pieces |