missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters

Product Code. 15317938
Click to view available options
No. of Places:
1
2
4
Electrical Requirements:
200/240 V, 50/60 Hz (CN, EU, UK)
This item is not returnable. View return policy
This item is not returnable. View return policy

Requires

Unit must be ordered with block(s)

Interchangeable modular blocks, configurable for a range of applications, increase lab versatility.

Fisherbrand™ Isotemp Digital Dry Baths/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs.

Fisherbrand™ Isotemp Digital Dry Baths/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs. Designed for basic to advanced applications with a maximum temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.

  • Digital controls and display of time and temperature
  • Advanced internal temperature sensing probe for outstanding temperature accuracy and control
  • Precise temperature control with PID circuit
  • Temperature calibration enables to offset the temperature to desired value
  • Timer allows the user to accurately monitor the heating time
  • Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
  • Powder-coated steel body construction ensures durability
  • Built-in over-temperature protection device warrants sample and user safety
  • Temperature deviation adjustment

Applications:

Sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation

Warranty

2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, cULus, RoHS
Controller Type PID
Material Powder-coated Steel
No. of Places 4
Plug Type CN, EU, UK
Relative Humidity <80%
Temperature Range (Metric) Ambient +5° to 130°C
Type Dry Bath/Block
Fuse 250V 3.15A
Heat Up Time ≤ 20 min. 30° to 130°C
Includes Dry bath, CN, EU and UK plugs, block lifter
Temperature Accuracy ≤ ± 0.5°C
Temperature Uniformity ≤ ± 1°C
Timer 0 to 99:59 min or continuous
Warranty 2 Year, Replacement
Dimensions (L x W x H) 17.7 x 7.9 x 3.9 in. (450 x 200 x 100 mm)
Length (English) 17.7 in.
Length (Metric) 450 mm
Width (English) 7.9 in.
Height (English) 3.9 in.
Height (Metric) 100 mm
Weight (English) 10.36 lb.
Weight (Metric) 4.7 kg
Electrical Requirements 200/240 V, 50/60 Hz (CN, EU, UK)
Voltage 200/240 V
Frequency 50/60 Hz
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'