missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Isotemp™ Gable Covers for Water Baths

Reduce moisture loss and water accumulation on the bench.

Brand:  Fisherbrand™ 1546235Q

 View more versions of this product

Product Code. 15197843

  • 352.00 € / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Stainless steel gable covers for water baths. Reduce moisture loss and water accumulation on the bench

  • 1 year warranty
  • Material: Stainless steel

Services

One year

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Model 220, 228 Isotemp™ 20L and 28L water bath
Stainless Steel
1 Year
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.