Learn More
Fisherbrand™ Min./Max. Thermometer
A safe alternative to glass thermometers.
Brand:  Fisherbrand™ 98768-84
Product Code. 11892623
  • 23.50 € / Each
Edge
Explore more special offers
Description
Description
  • Water-resistant and shockproof
  • Stainless-steel probe is resistant to most acids, bases and solvents
  • Reading updates every second
  • Memory recall of highest and lowest temperature readings, over period of 5 hours
  • Silver-oxide battery included
  • Wide temperature range
Specifications
Specifications
Silver Oxide battery
0.1 From −20 to +200°C
200°C
-10°C
-10°C to +200°C
Stainless Steel Probe
392°F
14°F
14°F to 392°F