Learn More
Fisherbrand™ MultiTube Vortexers: Post Extension Kit
Post extension kit for Fisherbrand™ MultiTube Vortexers.
Brand:  Fisherbrand™ 80862908
Product Code. 15539014
  • 180.00 € / Each
Estimated Shipment: 02-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with Fisher Scientific MultiTube Vortexers
  • Post extension kit adds 6 in. (15.2cm) to post to accommodate tubes up to 25.4cm (10 in.)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Post Extension Kit
Multitube vortexer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ MultiTube Vortexers: Post Extension Kit > Post Extension Kit