missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Probes for Model 50 and 120 Sonic Dismembrator

Image shows (left to right): 4423, 4422, 4435.

Brand:  Fisherbrand™ FB4435

295.17 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 12931181

  • 437.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

These Fisher Scientific™ Probes are designed for use with Model 50 and Model 120 Sonic Dismembrators.

  • 1/4 in. Probe for Model 50/120 Sonic Dismembrator
  • For processing 10 to 50mL volumes
Specifications

Specifications

120 μm maximum
10 to 50 mL
1/4 in.
6 mm
1 probe
Probe
0.25 in.
0.63 cm
Model 120 sonic dismembrator
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers