Learn More
Fisherbrand™ Replaceable Tip Probe for use with 500 and 700 watt Sonic Dismembrators
Microtips are thin, high intensity probes which are designed for processing small sample volumes. Microtips screw into the threaded end of the standard 1/2 in. probe (4220).
636.00 € - 949.00 €
Specifications
Product Type Probe
For Use With (Equipment) Model 505/705 Sonic dismembrator
View More Specs
Requires: Standard 1/2 in. probe (Cat. No. FB4220)
Products 6
Product Code Brand Diameter (Metric) Tip Volume Range (Metric) Includes Price Quantity & Availability  
Product Code Brand Diameter (Metric) Tip Volume Range (Metric) Includes Price Quantity & Availability  
12921171
View accessory products View Documents
Fisherbrand™
FB4220
13 mm 20 to 250 mL Standard 1/2 in. Probe
949.00 €
Each
Estimated Shipment: 03-07-2025
to see stock.
Add to basket
12971161
View accessory products View Documents
Fisherbrand™
FB4207
19 mm 50 to 500 mL Replacement tip
923.00 €
Each
Estimated Shipment: 03-07-2025
to see stock.
Add to basket
12981161
View Documents
Fisherbrand™
FB4208
19 mm 50 to 500 mL Probe
636.00 €
Each
Estimated Shipment: 03-07-2025
to see stock.
Add to basket
12991161
View Documents
Fisherbrand™
FB4209
25 mm 100 mL to 1 L Probe
641.00 €
Each
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
12901171
View accessory products View Documents
Fisherbrand™
FB4210
25 mm 100 mL to 1 L Replacement tip
641.00 €
Each
Estimated Shipment: 03-07-2025
to see stock.
Add to basket
12911171
View Documents
Fisherbrand™
FB4219
13 mm 20 to 250 mL Probe
672.00 €
Each
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
Description
Description
These Fisher Scientific™ Probes are designed for use with 500 and 700 watt Sonic Dismembrators.
  • Available in diameters ranging from 1/16 in. to 1 in.
  • Capacity options of 0.2-5mL to 100-1000mL
  • Replaceable tip, solid and microtip probes
Specifications
Specifications
Probe
Model 505/705 Sonic dismembrator
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Replaceable Tip Probe for use with 500 and 700 watt Sonic Dismembrators