missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Replacement Converter for Model 120 Sonic Dismembrator

Replacement 20kHz Converter for Model 120 Sonic Dismembrator

Brand:  Fisherbrand™ CL18

Product Code. 12931191

  • 1025.00 € / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • 20kHz Converter
  • For use with Model 120 Sonic Dismembrator
Specifications

Specifications

Replacement Converter
20 kHz Converter
Model 120 sonic dismembrator
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers