missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Replacement Converter for Model 505 and 705 Sonic Dismembrator
Replacement 20kHz Converter for Model 505/705 Sonic Dismembrator
Brand: Fisherbrand™ FBCL334
This item is not returnable.
View return policy
Description
- 20kHz Converter
- For use with Model 505 and 705 Sonic Dismembrator
Specifications
Replacement Converter | |
20 kHz Converter | |
Model 505 and 705 sonic dismembrator | |
1 Ea. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Replacement Converter for Model 505 and 705 Sonic Dismembrator > Replacement converter for 505 and 705 Sonic Dismembrators
Spot an opportunity for improvement?Share a Content Correction