Learn More
Fisherbrand™ Disposable Base Molds
For use in the preparation of pathology specimens
Brand:  Fisherbrand™ 22-363-554
 View more versions of this product
Product Code. 18405902
  • 23.63 € / Pack of 500
Edge
Explore more special offers
Description
Description
For use in the preparation of pathology specimens
  • Available in common base mold sizes; can be used with the same styles and types of cassettes and embedding rings
  • Molds are made from specially formulated plastic, offering excellent thermal exchange
  • A smooth interior finish and round corners facilitate specimen removal
Specifications
Specifications
Base Mold
2.4 cm
0.94 in.
0.5 cm
0.94 x 0.94 x 0.19 in. (2.4 x 2.4 x 0.5 cm)
Can be use with same styles and types of cassettes and embedding rings and provides offering excellent thermal exchange
Plastic
2.4 cm
0.94 in.
0.19 in.
Single-use
500/Pk.