Learn More
Fisherbrand™ Traceable™ 5mL Vaccine Bottle Probe
Accessory for Radio-Signal Remote Thermometer
Brand:  Fisherbrand™ 4618
Product Code. 11769755
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The Fisher Scientific™ Traceable™ 5mL Vaccine Bottle Probe monitors products subjected to door openings in freezers, incubators, and refrigerators
  • Mimics vaccine temperature
  • Fits in vaccine tray
  • Filled with patented nontoxic glycol solution for temperature buffering
Specifications
Specifications
Probe
Radio signal remote thermometer