missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Digital Thermometers with Short Sensors

Fisherbrand™ Traceable™ Digital Thermometers are ideal for monitoring liquids, air/gas or semisolids.

Brand:  Fisherbrand™ 90028-97

 View more versions of this product

Product Code. 11709765

  • 68.30 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Description

Description

Choose from models with a variety of features including MAX/MIN function to record the maximum and minimum temperature in a certain time period; visual and audible alarms and multiple displays of recorded temperatures.

  • Select the correct temperature to use in water baths, refrigerators, incubators and tanks

Certifications

Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™ indicates traceability to standards provided by NIST™.

Specifications

Specifications

DNV, ISO 17025, A2LA
4 x 4 x 0.62 in. (10.2 x 10.1 x 1.5 cm)
0.62 in.
Self adhesive wall mount and flip open lab bench stand
0.68 in.
91.44 cm
ABS Plastic
0.25 oz.
4 in.
4 in.
70°C
-50°C
-50°C to +70°C
Digital LCD
0.2 in.
Measuring temperatures of solutions in water baths, refrigerators, incubators and tanks
1.5 cm
3 ft.
102 mm
1.7 cm
0.1°
7 g
Digital Thermometer: Celsius Scale
158°F
-58°F
N/A
±1°C
10.1 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.