Learn More
Fisherbrand™ Traceable™ Digital Thermometers with Short Sensors
Fisherbrand™ Traceable™ Digital Thermometers are ideal for monitoring liquids, air/gas or semisolids.
Brand:  Fisherbrand™ 90008-16
 View more versions of this product
Product Code. 11799735
  • 108.00 € / Each
Edge
Explore more special offers
Description
Description
Choose from models with a variety of features including MAX/MIN function to record the maximum and minimum temperature in a certain time period; visual and audible alarms and multiple displays of recorded temperatures.
  • Select the correct temperature to use in water baths, refrigerators, incubators and tanks
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™ indicates traceability to standards provided by NIST™.
Specifications
Specifications
ISO 17025, A2LA, NIST
7 x 10.8 x 1.9 cm (2.75W x 4.25L x 0.75 in. D)
Sensor, AAA alkaline battery, flip open stand, wall mount, magnet and hook and loop mounting tape
0.75 in.
305 cm
1°C
113.3 g
158°F
-58°F
-58°F to +158°F
±1°C
0.31 in.
Monitoring liquids, air/gas or semisolids in water baths, refrigerators, freezers, incubators and tanks
10 ft.
70 mm
1.9 cm
4 oz.
Digital Thermometer: Dual Scale
70°C
-50°C
-50°C to +70°C
Digital LCD