Learn More
Fisherbrand™ Traceable™ Logger-Trac™ Temperature Datalogger Accessories
Accessories for Traceable™ Logger-Trac™ Temperature Datalogger.
Brand:  Fisherbrand™ 61011-25
 View more versions of this product
Product Code. 15338764
  • 86.55 € / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Accessories for Traceable™ Logger-Trac™ Temperature Datalogger.
15-081-145
  • Waterproof paper sleeves
  • For protecting temperature probe sensor
  • Transparent
15-081-148
  • USB standard A to Mini B, IC-9P or USB to serial converter
  • USB Version 2.0, ELV and RoHS compliant
  • Cable length: 1.5 m
  • Multiple cradles allow for batch programming up to 8 units
Specifications
Specifications
Waterproof Paper Sleeves
Paper
Fisherbrand Traceable Logger-Trac Temperature Datalogger
1.5 m
RoHS compliant
Transfer recorded information in dataloggers to computer
5 ft.