Learn More
Fisherbrand™ Traceable™ Stick-It Mini-Clock
Traceable™ Stick-It Mini-Clock is ideal for individual work areas with easily read bright hands and white sweeping second hand.
Brand:  Fisherbrand™ 90010-71
Product Code. 11745833
  • 23.25 € / Each
Explore more special offers
Description
Description
Traceable™ Stick-It Mini-Clock is ideal for individual work areas with easily read bright hands and white sweeping second hand.
  • Precise quartz-crystal accuracy is 0.013%
  • O-ring sealed metal case is water-resistant/dustproof
  • Flat bottom permits standing on shelf, desk, or bench
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO™ 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Specifications
Specifications
Mini Clock
1 sec.
0.3 in.
1.6 in.
1 oz.
Velcro™, double backed tape, 2 Years battery and traceable certificates
1 Ea.
0.013%
Rugged Metal
0.76 cm
4.06 cm
0.028 kg
Perfect for individual work areas with easily read bright hands and white sweeping second hand
ISO 17025, A2LA, NIST